missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Human/Mouse/Rat Thymosin-beta4 Recombinant Protein, PeproTech®

Product Code. 17839902
Click to view available options
Quantity:
100 μg
2 x 500 μg
20 μg
250 μg
500 μg
Unit Size:
100µg
1mg
20µg
250µg
500µg
Pack of 2
This item is not returnable. View return policy

Product Code. 17839902

Brand: Gibco™ 14014100UG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

140-14-1MG will be provided as 2 x 500 μg (140-14-500UG). Recombinant Human/Murine/Rat Thymosin-beta4 is a 5.2 kDa glycoprotein containing 45 amino acid residues. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

Thymosin-beta4 is a small, actin-sequestering protein belonging to the thymosin-beta family that is found at high concentrations within the spleen, thymus, and peritoneal macrophages, where it is most notably responsible for the organization of cytoskeletal structure. In mammalian tissues, this protein acts as a modulator for the polymerization/depolymerization of actin through the formation of a 1:1 complex with the monomer G (globular)-actin, and inhibits actin's polymerization to form F (filamentous) actin, which together with other proteins binds microfilaments to construct the cytoskeleton. Commonly found at significant quantities within the brain, lungs, liver, kidneys, testes, and heart, thymosin-beta4 has also been shown to be synthesized by cells unrelated to the reticuloendothelial system, such as myoblasts and fibroblasts, and expressed at irregular levels by several hemopoietic cell lines, malignant lymphoid cells and myeloma cells. In addition to regulating actin polymerization, research has also found thymosin-beta4 to stimulate the secretion of hypothalamic luteinizing hormone-releasing hormone and luteinizing hormone, inhibit the migration of peritoneal macrophages, induce phenotypic changes in T cell lines during early host defense mechanisms, and inhibit the progression of hematopoietic pluripotent stem cells into the S-phase.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P20065, P62328, P62329
For Use With (Application) Functional Assay
Formulation protein with no preservative
Gene ID (Entrez) 19241, 7114, 81814
Molecular Weight (g/mol) 5.2 kDa
Name Human/Mouse/Rat Thymosin-beta4
Quantity 100 μg
Source E. coli
Regulatory Status RUO
Endotoxin Concentration <1 EU/ μg
Gene Alias Fx; Hematopoietic system regulatory peptide; prothymosin beta 4; prothymosin beta-4; Ptmb4; Seraspenide; T beta 4; t beta-4; Tb4; TB4X; TB4Xbeta 4; Thyb4; thymosin; thymosin beta 4; thymosin beta 4, X-linked; thymosin beta-4; thymosin, beta 4, x chromosome; Tmsb4; Tmsb4x
Common Name TMSB4X
Gene Symbol TMSB4X
Biological Activity Pretreatment of primary lung fibroblasts with recombinant Thymosin-beta4, using a concentration of 0.5 - 10 ug/ml, produces a protective effect against hydrogen peroxide induced cell death.
Conjugate Unconjugated
Recombinant Recombinant
Sequence RMSDKPDMAE IEKFDKSKLK KTETQEKNPL PSKETIEQEK QAGES
Content And Storage -20°C
Expression System E. coli
Form Lyophilized
Purity or Quality Grade ≥ 95% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.