missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse/Rat Thymosin beta 10 Antibody, R&D Systems™

Sheep Polyclonal Antibody
Brand: R&D Systems AF6429
This item is not returnable.
View return policy
Description
Thymosin beta 10 Polyclonal specifically detects Thymosin beta 10 in Human, Mouse, Rat samples. It is validated for Western Blot.
Specifications
| Thymosin beta 10 | |
| Polyclonal | |
| Unconjugated | |
| P63313 | |
| TMSB10 | |
| E. coli-derived recombinant human Thymosin beta 10 Ala2-Ser44 Accession # P63313 | |
| 100 μg | |
| Primary | |
| Detects human Thymosin beta 10 in direct ELISAs and Western blots. In direct ELISAs, less than 1% cross-reactivity with recombinant human (rh) Thymosin beta 4 and rhThymosin beta 16 is observed. | |
| Human, Mouse, Rat | |
| IgG |
| Western Blot | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| MIG12, migration-inducing gene 12, migration-inducing protein 12, PTMB10, TB10, THYB10, thymosin beta 10, thymosin beta-10, TMSB10 | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 9168 | |
| Sterile PBS to a final concentration of 0.2 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction