missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse/Rat ST3 beta-Gal alpha-2,3-Sialyltransferase 2/ST3GAL2 Antibody, R&D Systems™

Sheep Polyclonal Antibody
Brand: R&D Systems AF7275-SP
This item is not returnable.
View return policy
Description
ST3 beta-Gal alpha-2,3-Sialyltransferase 2/ST3GAL2 Polyclonal specifically detects ST3 beta-Gal alpha-2,3-Sialyltransferase 2/ST3GAL2 in Human, Mouse, Rat samples. It is validated for Western Blot.
Specifications
| ST3 beta-Gal alpha-2,3-Sialyltransferase 2/ST3GAL2 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| Alpha 2,3-ST 2, Beta-galactoside alpha-2,3-sialyltransferase 2, beta-galactoside alpha-2,3-sialytransferase, CMP-N-acetylneuraminate-beta-galactosamide-alpha-2,3-sialyltransferase 2, EC 2.4.99.4, EC 2.4.99.7, Gal-beta-1,3-GalNAc-alpha-2,3-sialyltransferase, Gal-NAc6S, Sialyltransferase 4B, sialyltransferase 4B (beta-galactosidase alpha-2,3-sialytransferase), SIAT4B, SIAT4-B, ST3 beta-galactoside alpha-2,3-sialyltransferase 2, ST3GalA.2, ST3GalA.2ST3Gal II, ST3GALII | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 6483 | |
| Sterile PBS to a final concentration of 0.2 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot | |
| LYOPH | |
| Western Blot 0.5 ug/mL | |
| NP_008858 | |
| ST3GAL2 | |
| Chinese hamster ovary cell line CHO-derived recombinant human ST3 beta-Gal alpha-2,3-Sialyltransferase 2/ST3GAL2 Pro52-Gln350 Accession # NP_008858 | |
| 25 μg | |
| Primary | |
| Detects human, mouse and rat ST3 beta-Gal alpha-2,3-Sialyltransferase 2/ST3GAL2 in Western blots. In direct ELISAs, less than 1% cross-reactivity with recombinant human (rh) ST6GALNAC4 and rhST3GAL1 is observed. | |
| Human, Mouse, Rat | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction