missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse/Rat Sortilin Antibody, R&D Systems™
Mouse Monoclonal Antibody
Brand: R&D Systems MAB3154
This item is not returnable.
View return policy
Description
Sortilin Monoclonal specifically detects Sortilin in Human, Mouse, Rat samples. It is validated for Western Blot, Simple Western, Knockout Validated.
Specifications
| Sortilin | |
| Monoclonal | |
| LYOPH | |
| Western Blot 0.25-1 ug/mL, Simple Western 10 ug/mL, Knockout Validated | |
| Q99523 | |
| SORT1 | |
| Mouse myeloma cell line NS0-derived recombinant human Sortilin Ser78-Asn755 Accession # Q99523 | |
| 100 μg | |
| Primary | |
| Detects human, mouse, and rat Sortilin in Western blots. | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, KnockDown | |
| 334708 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| 100 kDa NT receptor, Glycoprotein 95, Gp95, Gp95LDLCQ6, Neurotensin receptor 3, NT3NTR3, Ntr3, SORT1, sortilin, sortilin 1 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 6272 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction