missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse/Rat SOD2/Mn-SOD Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems AF3419-SP
This item is not returnable.
View return policy
Description
SOD2/Mn-SOD Polyclonal specifically detects SOD2/Mn-SOD in Human, Mouse, Rat samples. It is validated for Western Blot, Simple Western.
Specifications
| SOD2/Mn-SOD | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| EC 1.15.1.1, indophenoloxidase B, IPOB, IPO-B, manganese-containing superoxide dismutase, mangano-superoxide dismutase, Mn SOD, Mn superoxide dismutase, MnSOD, MVCD6, SOD, Mitochodrial, superoxide dismutase [Mn], mitochondrial, superoxide dismutase 2, mitochondrial | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 6648 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot | |
| LYOPH | |
| Western Blot 1 ug/mL, Simple Western 10 ug/mL | |
| P04179 | |
| SOD2 | |
| E. coli-derived recombinant human SOD2 Lys25-Lys222 Accession # P04179 | |
| 25 μg | |
| Primary | |
| Detects human, mouse and rat SOD2 in Western blots. | |
| Human, Mouse, Rat | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction