missing translation for 'onlineSavingsMsg'
Learn More

Proteintech™ Human/Mouse/Rat SNAP25 ELISA Kit

Product Code. 16851456 Shop All Proteintech Products
Click to view available options
Target Species:
Human, Mouse, Rat
Unit Size:
96 tests
This item is not returnable. View return policy

Product Code. 16851456

Brand: Proteintech™ KE0003196T

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

KE00031 is a solid phase sandwich Enzyme Linked-Immuno-Sorbent Assay (Sandwich ELISA). The SNAP25 ELISA kit is to be used to detect and quantify protein levels of endogenous SNAP25.

  • Synaptic vesicle membrane docking and fusion is mediated by soluble N-ethylmaleimide-sensitive factor attachment protein receptors (SNARE) complex which is located on the vesicle membrane (v-SNAREs) and the target membrane (t-SNAREs). Other than VAMP2 and STX1A, SNAP25 is another key component of SNARE core complex, and is known to be involved in regulating neurotransmitter release.
  • Its palmitoylation domain is located in the middle of the molecule that contains four cysteine residues and mutation of the cysteines abolishes palmitoylation and its membrane binding activity. As an important presynaptic plasma membrane protein, SNAP25 has been linked to memory and learning by its effect on long term potentiation in the hippocampus, thus playing a critical role in the synaptic function of specific neuronal systems.
  • It has been reported that SNAP25 levels were were significantly decreased in brain tissue homogenates of patients with later stages of Alzheimer's disease compared with the controls, while the levels of SNAP25 were significantly increased in cerebrospinal fluid (CSF) of the group with an Alzheimer's disease biomarker profile than in the group with a control biomarker profile.
  • The assay recognizes human SNAP25. An antibody specific for SNAP25 has been pre-coated onto the microwells. The SNAP25 protein in samples is captured by the coated antibody after incubation.
  • Following extensive washing, another antibody specific for SNAP25 is added to detect the captured SNAP25 protein. For signal development, horseradish peroxidase (HRP)-conjugated antibody is added, followed by Tetramethyl-benzidine (TMB) reagent.
  • Solution containing sulfuric acid is used to stop color development and the color intensity which is proportional to the quantity of bound protein is measurable at 450 nm with the correction wavelength set at 630 nm.

Specifications

Accession Number NP_003072
Assay Range 15.6 to 1000 pg/mL
Assay Sensitivity 6 pg/mL
Certifications/Compliance ISO 13485
Detection Method Sandwich ELISA, Double Antibody
Immunoassay Kit Format Coated ELISA Kit
Sample Type Plasma, Serum
Substrate DAB
Target Species Human, Mouse, Rat
Gene ID (Entrez) 6616
Host Species Human/Mouse/Rat
Immunoassay Kit Grade Solid
Interassay CV <10%
Intraassay CV <10%
Shipping Condition Blue ice
Storage Requirements -20°C to +4°C
Total Assay Time 3.5 hr.
UniProt ID P60880
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 96TEST Human/Mouse/Rat SNAP25 ELISA Kit
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P202-Do not handle until all safety precautions have been read and understood.
  • P234-Keep only in original container.
  • P264-Wash skin thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P330-Rinse mouth.
  • P362-Take off contaminated clothing.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : Sulphuric Acid, Tetramethylbenzidine
Corrección del contenido de un producto

Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.

Título del producto

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.

Gracias por ayudarnos a mejorar nuestra web. Su comentario ha sido enviado