missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse/Rat SMURF2 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB6916
This item is not returnable.
View return policy
Description
SMURF2 Monoclonal specifically detects SMURF2 in Human, Mouse, Rat samples. It is validated for Western Blot.
Specifications
| SMURF2 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| Q9HAU4 | |
| SMURF2 | |
| E. coli-derived recombinant human SMURF2 Met490-Glu748 Accession # Q9HAU4 | |
| 100 μg | |
| Primary | |
| Detects human SMURF2 in direct ELISAs and Western blots. In Western blots, no cross-reactivity with recombinant human SMURF1 is observed. | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot | |
| 690638 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| DKFZp686F0270, E3 ubiquitin ligase SMURF2, E3 ubiquitin-protein ligase SMURF2, EC 6.3.2, EC 6.3.2.-, hSMURF2, MGC138150, SMAD specific E3 ubiquitin protein ligase 2, SMAD ubiquitination regulatory factor 2, SMAD-specific E3 ubiquitin-protein ligase 2 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 64750 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction