missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse/Rat SMC1 Antibody, R&D Systems™
Goat Polyclonal Antibody
Brand: R&D Systems AF3294
This item is not returnable.
View return policy
Description
SMC1 Polyclonal specifically detects SMC1 in Human, Mouse, Rat samples. It is validated for Western Blot.
Specifications
| SMC1 | |
| Polyclonal | |
| Unconjugated | |
| Q14683.2 | |
| SMC1A | |
| E. coli-derived recombinant human SMC1 Met837-Gln1233 Accession # Q14683.2 | |
| 100 μg | |
| Primary | |
| Detects human, mouse, and rat SMC1 in Western blots. | |
| Human, Mouse, Rat | |
| IgG |
| Western Blot | |
| LYOPH | |
| Western Blot 0.5 ug/mL | |
| CDLS2, DXS423ESMC1 structural maintenance of chromosomes 1-like 1 (yeast), KIAA0178DKFZp686L19178, SB1.8, SB1.8MGC138332, segregation of mitotic chromosomes 1, SMC protein 1A, SMC1 (structural maintenance of chromosomes 1, yeast)-like 1, SMC1 structural maintenance of chromosomes 1-like 1, SMC1A, SMC-1A, SMC-1-alpha, SMC1L1, SMC1SMC1alpha, SMCB, structural maintenance of chromosomes 1A, structural maintenance of chromosomes protein 1A | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 8243 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction