missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse/Rat SF20/MYDGF Antibody, R&D Systems™

Rabbit Monoclonal Antibody
Brand: R&D Systems MAB1104-100
This item is not returnable.
View return policy
Description
SF20/MYDGF Monoclonal specifically detects SF20/MYDGF in Human, Mouse, Rat samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Intracellular Staining by Flow Cytometry, Immunocytochemistry, CyTOF-ready.
Specifications
| SF20/MYDGF | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| C19orf10, chromosome 19 open reading frame 10, EUROIMAGE1875335, IL25, IL27, IL27w, interleukin 27 working designation, Interleukin-25, Ly6elg, MYDGF, myeloid-derived growth factor, R33729_1, SF20, Stromal cell-derived growth factor SF20, UPF0556 protein C19orf10 | |
| Rabbit | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| 56005 | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Immunohistochemistry, Flow Cytometry, Immunocytochemistry, CyTOF | |
| 2562A | |
| Western Blot 1 ug/mL, Simple Western 20 ug/mL, Immunohistochemistry 0.3-25 ug/mL, Intracellular Staining by Flow Cytometry 0.25 ug/10^6 cells, Immunocytochemistry 3-25 ug/mL, CyTOF-ready | |
| Q9CPT4 | |
| MYDGF | |
| Mouse myeloma cell line NS0-derived mouse SF20/MYDGF, Val25-Leu166, Accession # Q9CPT4 | |
| 100 μg | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction