missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse/Rat SEC13 Antibody, R&D Systems™

Rabbit Monoclonal Antibody
£156.00 - £369.00
Specifications
| Antigen | SEC13 |
|---|---|
| Clone | 1280C |
| Dilution | Western Blot 0.1 ug/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18766598
![]() |
R&D Systems
MAB90552-SP |
25 μg |
£156.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18759208
![]() |
R&D Systems
MAB90552-100 |
100 μg |
£369.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SEC13 Monoclonal specifically detects SEC13 in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
| SEC13 | |
| Western Blot 0.1 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Supplied as a solution in PBS containing BSA, Glycerol and Sodium Azide. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with Sodium Azide | |
| D3S1231E, D3S1231ESEC13L1npp-20, Npp-20, protein SEC13 homolog, SEC13 (S. cerevisiae)-like 1, SEC13 homolog (S. cerevisiae), SEC13L1, SEC13-like 1 isoform, SEC13-like protein 1, SEC13R, SEC13-related protein, SEC13RSEC13-like 1 (S. cerevisiae) | |
| E. coli-derived recombinant human SEC13 Gly123-Gln322 Accession # P55735 | |
| Primary | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| 1280C | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| P55735 | |
| 6396 | |
| IgG | |
| Protein A or G purified from hybridoma culture supernatant | |
| Detects human SEC13 in direct ELISAs and Western blots. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title