missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse/Rat ROCK1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB4590-SP
This item is not returnable.
View return policy
Description
ROCK1 Monoclonal specifically detects ROCK1 in Human, Mouse, Rat samples. It is validated for Western Blot.
Specifications
| ROCK1 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 2 ug/mL | |
| Q13464 | |
| ROCK1 | |
| E. coli-derived recombinant human ROCK1 Ala629-Thr829 Accession # Q13464 | |
| 25 μg | |
| Primary | |
| Detects human ROCK1 in direct ELISAs and Western blots. In Western blots, no cross-reactivity with recombinant human ROCK2 is observed. | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot | |
| 701118 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| EC 2.7.11, EC 2.7.11.1, MGC131603, MGC43611, p160 ROCK-1, p160ROCK, p160-ROCK, PRO0435, Renal carcinoma antigen NY-REN-35, Rho kinase, rho-associated protein kinase 1, Rho-associated, coiled-coil containing protein kinase 1, Rho-associated, coiled-coil-containing protein kinase 1, ROK beta | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 6093 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction