Learn More
Human/Mouse/Rat Ribosomal Protein S6/RPS6 Antibody, R&D Systems™
Sheep Polyclonal Antibody
Brand:  R&D Systems AF5436
Product Code. 18012852
  • £461.00 / 100µg
Description
Ribosomal Protein S6/RPS6 Polyclonal specifically detects Ribosomal Protein S6/RPS6 in Human, Mouse, Rat samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Ribosomal Protein S6/RPS6
Polyclonal
Unconjugated
P62753
RPS6
E. coli-derived recombinant human Ribosomal Protein S6 Met1-Lys249 Accession # P62753
100 μg
Primary
Detects human, mouse and rat Ribosomal Protein S6 in Western blots.
Human, Mouse, Rat
IgG
Western Blot
LYOPH
Western Blot 1 ug/mL
Phosphoprotein NP33, 40S ribosomal protein S6, pS6, ribosomal protein S6, RPS6, S6
Sheep
Affinity Purified
RUO
6194
Sterile PBS to a final concentration of 0.2 mg/mL.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.