missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse/Rat RalA/RalB Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems AF3204-SP
This item is not returnable.
View return policy
Description
RalA/RalB Polyclonal specifically detects RalA/RalB in Human, Mouse, Rat samples. It is validated for Western Blot.
Specifications
| RalA/RalB | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| MGC48949, RAL, Ras family small GTP binding protein RALA, Rasl1, RAS-like protein A, ras-related protein Ral-A, v-ral simian leukemia viral oncogene homolog A (ras related) | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 5898 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| P11234 | |
| RALA | |
| E. coli-derived recombinant human RalB Met1-Leu206 Accession # P11234 | |
| 25 μg | |
| Primary | |
| Detects human, mouse, and rat RalA/RalB in Western blots. In Western blots, less than 1% cross-reactivity with recombinant human H-Ras, N-Ras, or K-Ras is observed. | |
| Human, Mouse, Rat | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction