missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse/Rat PTP-MEG2 Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems AF2668
This item is not returnable.
View return policy
Description
PTP-MEG2/PTPN9 Polyclonal specifically detects PTP-MEG2/PTPN9 in Human, Mouse, Rat samples. It is validated for Western Blot.
Specifications
| PTP-MEG2/PTPN9 | |
| Polyclonal | |
| Western Blot 1 ug/mL | |
| P43378 | |
| PTPN9 | |
| E. coli-derived recombinant human PTP-MEG2 Met1-Gln593 Accession # P43378 | |
| 100 μg | |
| Primary | |
| Detects human, mouse, and rat PTP-MEG2 in Western blots. | |
| Human, Mouse, Rat | |
| IgG |
| Western Blot | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| EC 3.1.3.48, MEG2, protein tyrosine phosphatase, non-receptor type 9, Protein-tyrosine phosphatase MEG2, PTPase MEG2, PTPase-MEG2, PTPMEG2, PTPN9, tyrosine-protein phosphatase non-receptor type 9 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 5780 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction