missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse/Rat PTP-MEG2 Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 5 publications
Brand: R&D Systems MAB2668-SP
This item is not returnable.
View return policy
Description
PTP-MEG2/PTPN9 Monoclonal specifically detects PTP-MEG2/PTPN9 in Human, Mouse, Rat samples. It is validated for Western Blot.
Specifications
| PTP-MEG2/PTPN9 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| P43378 | |
| PTPN9 | |
| E. coli-derived recombinant human PTP-MEG2 Glu2-Gln593 Accession # P43378 | |
| 25 μg | |
| Primary | |
| Detects human, mouse, and rat PTP-MEG2. | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Immunocytochemistry | |
| 291835 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| EC 3.1.3.48, MEG2, protein tyrosine phosphatase, non-receptor type 9, Protein-tyrosine phosphatase MEG2, PTPase MEG2, PTPase-MEG2, PTPMEG2, PTPN9, tyrosine-protein phosphatase non-receptor type 9 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 5780 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction