missing translation for 'onlineSavingsMsg'
Learn More

Human/Mouse/Rat PTP gamma/PTPRG Antibody, R&D Systems™
GREENER_CHOICE

Product Code. 18791369
Change view
Click to view available options
Quantity:
25 μg
Unit Size:
25µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18791369 25 μg 25µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18791369 Supplier R&D Systems Supplier No. MAB4608SP

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody has been used in 2 publications

PTP gamma/PTPRG Monoclonal specifically detects PTP gamma/PTPRG in Human, Mouse, Rat samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PTP gamma/PTPRG
Applications Western Blot
Classification Monoclonal
Clone 481505
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P23470
Gene Alias EC 3.1.3.48, H_RG317H01.1, HPTPG, protein tyrosine phosphatase, receptor type, G, protein tyrosine phosphatase, receptor type, gamma polypeptide, Protein-tyrosine phosphatase gamma, PTPG, PTPGprotein tyrosine phosphatase gamma, PTPRG, receptor type protein tyrosine phosphatase gamma, receptor tyrosine phosphatase gamma, receptor-type protein phosphatase gamma, receptor-type tyrosine-protein phosphatase gamma, RPTPG, R-PTP-gamma
Gene Symbols PTPRG
Host Species Mouse
Immunogen E. coli-derived recombinant human PTP gamma/PTPRG Ser111-Ala541 Accession # P23470
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5793
Test Specificity Detects human, mouse, and rat PTP gamma/PTPRG in Western blots. In Western blots, no cross-reactivity with recombinant human PTPRM, PTPRK, PTPRS, LAR, or DEP1 is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human, Mouse, Rat
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.