missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse/Rat Properdin Antibody, R&D Systems™

Sheep Polyclonal Antibody
Brand: R&D Systems AF8216-SP
This item is not returnable.
View return policy
Description
Properdin Polyclonal specifically detects Properdin in Human, Mouse, Rat samples. It is validated for Western Blot.
Specifications
| Properdin | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| BFD, CFP, Complement Factor P, complement factor properdin, PFC, PFCcomplement, PFD, PROPERDIN | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 5199 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| P27918 | |
| CFP | |
| Human embryonic kidney cell line HEK293-derived human Properdin Asp28-Leu469 Accession # P27918 | |
| 25 μg | |
| Primary | |
| Detects human, mouse, and rat Properdin in direct ELISAs and Western blots. | |
| Human, Mouse, Rat | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction