missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse/Rat Plexin A2 APC-conjugated Antibody, R&D Systems™
Goat Polyclonal Antibody
Brand: R&D Systems FAB5486A
This item is not returnable.
View return policy
Description
Plexin A2 Polyclonal specifically detects Plexin A2 in Human, Mouse, Rat samples. It is validated for Flow Cytometry.
Specifications
| Plexin A2 | |
| Polyclonal | |
| Flow Cytometry 10 uL/10^6 cells | |
| FLJ11751, FLJ30634, KIAA0463plexin 2, OCT, OCTSemaphorin receptor OCT, plexin A2, PLXN2, PLXN2plexin-A2, PLXNA2, transmembrane protein OCT | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 5362 | |
| Human, Mouse, Rat | |
| IgG |
| Flow Cytometry | |
| APC | |
| P70207 | |
| PLXNA2 | |
| Chinese hamster ovary cell line CHO-derived recombinant mouse Plexin A2 Met35-Pro1237 Accession # P70207 | |
| 100 Tests | |
| Primary | |
| Detects mouse and rat Plexin A2 in direct ELISAs and Western blots. In direct ELISAs, less than 1% cross-reactivity with recombinant mouse (rm) Plexin A1, rmPlexin A3, and recombinant human Plexin B1 is observed. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction