missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse/Rat PKC epsilon Antibody, R&D Systems™
Sheep Polyclonal Antibody has been used in 2 publications
Brand: R&D Systems AF5134
This item is not returnable.
View return policy
Description
PKC epsilon Polyclonal specifically detects PKC epsilon in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| PKC epsilon | |
| Polyclonal | |
| Unconjugated | |
| Q02156 | |
| PRKCE | |
| E. coli-derived recombinant human PKC epsilon Gln580-Pro737 Accession # Q02156 | |
| 100 μg | |
| Primary | |
| Detects human, mouse and rat PKC epsilon in Western blots. | |
| Human, Mouse, Rat | |
| IgG |
| Western Blot, Immunohistochemistry | |
| LYOPH | |
| Western Blot 1 ug/mL, Immunohistochemistry 5-15 ug/mL | |
| EC 2.7.11, EC 2.7.11.13, MGC125656, nPKC-epsilon, PKCE, PKCEMGC125657, PRKCE, protein kinase C epsilon type, protein kinase C, epsilon | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 5581 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction