missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse/Rat Phospho-JNK (T183/Y185) Antibody, R&D Systems™
Rabbit Polyclonal Antibody has been used in 12 publications
Brand: R&D Systems AF1205
This item is not returnable.
View return policy
Description
JNK1/2/3 Polyclonal specifically detects JNK1/2/3 in Human, Mouse, Rat samples. It is validated for Western Blot, Simple Western, Immunohistochemistry.
Specifications
| JNK1/2/3 (p Thr183, p Tyr185) | |
| Polyclonal | |
| Unconjugated | |
| c-Jun N-terminal kinase 1, EC 2.7.11, EC 2.7.11.24, JNK1 alpha protein kinase, JNK1 beta protein kinase, JNK1JNK1A2, JNK-46, JUN N-terminal kinase, MAP kinase 8, MAPK 8, MAPK10, MAPK8, MAPK9, mitogen-activated protein kinase 8, mitogen-activated protein kinase 8 isoform JNK1 alpha1, mitogen-activated protein kinase 8 isoform JNK1 beta2, PRKM8, PRKM8JNK, protein kinase JNK1, SAPK1, SAPK1 alpha, SAPK1 beta, SAPK1JNK21B1/2, Stress-activated protein kinase 1, Stress-activated protein kinase JNK1 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 5599 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Immunohistochemistry | |
| LYOPH | |
| Western Blot 0.5 ug/mL, Simple Western 5 ug/mL, Immunohistochemistry 5-15 ug/mL | |
| MAPK8IP3 | |
| Phosphopeptide containing human, rat, and mouse JNK1 T183/Y185 site | |
| 50 μg | |
| Primary | |
| Detects human, mouse and rat p46 and p54 JNK when dually phosphorylated at sites homologous to T183/Y185 of JNK1 and JNK2, and T221/Y223 of JNK3 in Western blots. | |
| Human, Mouse, Rat | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction