missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse/Rat Phospho-FRS2 (Y436) Antibody, R&D Systems™

Rabbit Polyclonal Antibody has been used in 6 publications
Brand: R&D Systems AF5126-SP
This item is not returnable.
View return policy
Description
FRS2 Polyclonal specifically detects FRS2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry.
Specifications
| FRS2 (p tyr436) | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| FRS2 | |
| Phosphopeptide containing human FRS2 Y436 site | |
| 25 μg | |
| Primary | |
| Detects human, mouse, and rat FRS2 isoforms when phosphorylated at Y436. | |
| Human, Mouse, Rat | |
| IgG |
| Western Blot, Immunocytochemistry | |
| LYOPH | |
| Western Blot 1 ug/mL, Immunocytochemistry 5-15 ug/mL | |
| FGFR signalling adaptor, FGFR-signaling adaptor SNT, fibroblast growth factor receptor substrate 2, FRS2A, FRS2alpha, SNT, SNT1, SNT-1FGFR substrate 2, Suc1-associated neurotrophic factor target 1 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 10818 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction