missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse/Rat Peroxiredoxin 1 Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 2 publications
Brand: R&D Systems AF3488
This item is not returnable.
View return policy
Description
Peroxiredoxin 1 Polyclonal specifically detects Peroxiredoxin 1 in Human, Mouse, Rat samples. It is validated for Western Blot, Simple Western.
Specifications
| Peroxiredoxin 1 | |
| Polyclonal | |
| Unconjugated | |
| Q06830 | |
| PRDX1 | |
| E. coli-derived recombinant human Peroxiredoxin 1 Met1-Lys199 Accession # Q06830 | |
| 100 μg | |
| Primary | |
| Detects human, mouse and rat Peroxiredoxin 1 in Western blots. In Western blots, no cross-reactivity with recombinant human Peroxiredoxin 2, 3, 5, or 6 is observed. | |
| Human, Mouse, Rat | |
| IgG |
| Western Blot | |
| LYOPH | |
| Western Blot 0.2 ug/mL, Simple Western 2 ug/mL | |
| EC 1.11.1, EC 1.11.1.15, Natural killer cell-enhancing factor A, natural killer-enhancing factor A, NKEFA, NKEF-A, PAG, PAGAMSP23, PAGB, peroxiredoxin 1, peroxiredoxin-1, PRDX1, proliferation-associated gene A, Proliferation-associated gene protein, PRX1, PRXI, TDPX2, Thioredoxin peroxidase 2, Thioredoxin-dependent peroxide reductase 2 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 5052 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction