missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse/Rat Peroxiredoxin 1 Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 2 publications
Brand: R&D Systems AF3488-SP
This item is not returnable.
View return policy
Description
Peroxiredoxin 1 Polyclonal specifically detects Peroxiredoxin 1 in Human, Mouse, Rat samples. It is validated for Western Blot, Simple Western.
Specifications
| Peroxiredoxin 1 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| EC 1.11.1, EC 1.11.1.15, Natural killer cell-enhancing factor A, natural killer-enhancing factor A, NKEFA, NKEF-A, PAG, PAGAMSP23, PAGB, peroxiredoxin 1, peroxiredoxin-1, PRDX1, proliferation-associated gene A, Proliferation-associated gene protein, PRX1, PRXI, TDPX2, Thioredoxin peroxidase 2, Thioredoxin-dependent peroxide reductase 2 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 5052 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot | |
| LYOPH | |
| Western Blot 0.2 ug/mL, Simple Western 2 ug/mL | |
| Q06830 | |
| PRDX1 | |
| E. coli-derived recombinant human Peroxiredoxin 1 Met1-Lys199 Accession # Q06830 | |
| 25 μg | |
| Primary | |
| Detects human, mouse and rat Peroxiredoxin 1 in Western blots. In Western blots, no cross-reactivity with recombinant human Peroxiredoxin 2, 3, 5, or 6 is observed. | |
| Human, Mouse, Rat | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction