missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse/Rat PBEF/Visfatin Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB40441-SP
This item is not returnable.
View return policy
Description
PBEF/Visfatin/NAMPT Monoclonal specifically detects PBEF/Visfatin/NAMPT in Human, Mouse, Rat samples. It is validated for Western Blot, Simple Western.
Specifications
| PBEF/Visfatin/NAMPT | |
| Monoclonal | |
| LYOPH | |
| Western Blot 0.5-1 ug/mL, Simple Western 5-50 ug/mL | |
| P43790 | |
| NAMPT | |
| E. coli-derived recombinant human PBEF/Visfatin Pro27-His491 Accession # P43790 | |
| 25 μg | |
| Primary | |
| Detects human PBEF/Visfatin in direct ELISAs and Western blots. | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot | |
| 882104 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| EC 2.4.2.12, MGC117256, NAmPRTase, NAMPT, nicotinamide phosphoribosyltransferase, NMPRTase, PBEF1, PBEF1110035O14Rik, PBEF1DKFZp666B131, Pre-B cell-enhancing factor, pre-B-cell colony enhancing factor 1, Pre-B-cell colony-enhancing factor 1, VF, VISFATIN | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 10135 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction