missing translation for 'onlineSavingsMsg'
Learn More

Human/Mouse/Rat Pannexin-1 Antibody, R&D Systems™

Product Code. 18198980
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18198980 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18198980 Supplier R&D Systems Supplier No. MAB7097

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody has been used in 4 publications

Pannexin-1 Monoclonal specifically detects Pannexin-1 in Human, Mouse, Rat samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Pannexin-1
Applications Western Blot
Classification Monoclonal
Clone 720505
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q96RD7
Gene Alias Innexin, MGC21309, MRS1, MRS1innexin, pannexin 1, pannexin-1, Pannexin1, PANX1, PX1, UNQ2529
Gene Symbols PANX1
Host Species Mouse
Immunogen E. coli-derived recombinant human Pannexin-1 Pro298-Asp376 Accession # Q96RD7
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 24145
Test Specificity Detects human Pannexin-1 in direct ELISAs and Western blots. In Western blots, no cross-reactivity with recombinant human Pannexin-2 or -3 is observed.
Reconstitution Sterile PBS to a final concentration of 0.5 mg/mL.
Target Species Human, Mouse, Rat
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.