Learn More
Human/Mouse/Rat p53 Agarose-conjugated Antibody, R&D Systems™
Goat Polyclonal Antibody
Brand:  R&D Systems GAF1355
Product Code. 18182631
  • £331.00 / 250µL
Description
p53 Polyclonal specifically detects p53 in Human, Mouse, Rat samples. It is validated for Immunoprecipitation.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
p53
Polyclonal
Immunoprecipitation
TP53
Affinity Purified
RUO
7157
Human, Mouse, Rat
IgG
Immunoprecipitation
Agarose
Antigen NY-CO-13, BCC7, FLJ92943, LFS1, LFS1TRP53, p53, p53 tumor suppressor, P53cellular tumor antigen p53, Phosphoprotein p53, TP53, transformation-related protein 53, TRP53, tumor protein p53, Tumor suppressor p53
Goat
250 μL
Primary
Sterile PBS to a final concentration of 0.2 mg/mL.
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.