missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse/Rat p38 gamma Antibody, R&D Systems™
Goat Polyclonal Antibody
Brand: R&D Systems AF1644
This item is not returnable.
View return policy
Description
p38 gamma/SAPK3 Polyclonal specifically detects p38 gamma/SAPK3 in Human, Mouse, Rat samples. It is validated for Western Blot.
Specifications
| p38 gamma/SAPK3 | |
| Polyclonal | |
| Unconjugated | |
| P53778 | |
| MAPK12 | |
| E. coli-derived recombinant human p38 gamma Met1-Leu367 Accession # P53778 | |
| 100 μg | |
| Primary | |
| Detects human, mouse, and rat p38 gamma in Western blots. The antibody is highly specific for p38 gamma and in Western blots, shows extremely weak cross-reactivity with recombinant p38 alpha , p38 beta , or p38 delta after prolonged exposure to film. | |
| Human, Mouse, Rat | |
| IgG |
| Western Blot | |
| LYOPH | |
| Western Blot 0.2 ug/mL | |
| EC 2.7.11, ERK3, ERK-6, ERK6MAPK 12, Extracellular signal-regulated kinase 6, MAP kinase 12, MAP kinase p38 gamma, MAPK12, mitogen-activated protein kinase 12, mitogen-activated protein kinase 3, Mitogen-activated protein kinase p38 gamma, P38GAMMA, PRKM12, SAPK-3, SAPK3EC 2.7.11.24, Stress-activated protein kinase 3 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 6300 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction