missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse/Rat p130Cas Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB5730-SP
This item is not returnable.
View return policy
Description
p130Cas/BCAR1 Monoclonal specifically detects p130Cas/BCAR1 in Human, Mouse, Rat samples. It is validated for Western Blot.
Specifications
| p130Cas/BCAR1 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| P56945 | |
| BCAR1 | |
| E. coli-derived recombinant human p130Cas Met66-Val412 Accession # P56945 | |
| 25 μg | |
| Primary | |
| Detects human p130Cas in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human Ajuba is observed. | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot | |
| 685718 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| BCAR1, breast cancer estrogen resistance 1, Cas scaffolding protein family member 1, CAS1, CASFLJ45059, CASS1, CASS1CRKASbreast cancer estrogen resistance protein 1, CRKAS, CRK-associated substrate, Crk-associated substrate p130Cas, FLJ12176, P130Cas | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 9564 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction