missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse/Rat Nrf1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB5306-SP
This item is not returnable.
View return policy
Description
Nrf1 Monoclonal specifically detects Nrf1 in Human, Mouse, Rat samples. It is validated for Western Blot.
Specifications
| Nrf1 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 2 ug/mL | |
| AAH10623 | |
| NRF1 | |
| E. coli-derived recombinant human Nrf1 Met1-Lys742 Accession # AAH10623 | |
| 25 μg | |
| Primary | |
| Detects endogenous human, mouse and rat Nrf1 at 85 kDa in Western blots. | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot | |
| 492413 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| Alpha palindromic-binding protein, Alpha-pal, EWG, HBZ17, LCR-F1, NFE2L1, NRF-1, nuclear respiratory factor 1, TCF11 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 4899 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction