missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse/Rat NFATC3 Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems AF5834
This item is not returnable.
View return policy
Description
NFATC3/NFAT4 Polyclonal specifically detects NFATC3/NFAT4 in Human, Mouse, Rat samples. It is validated for Western Blot.
Specifications
| NFATC3/NFAT4 | |
| Polyclonal | |
| Unconjugated | |
| Q12968 | |
| NFATC3 | |
| E. coli-derived recombinant human NFATC3 Asp900-Asp1035 Accession # Q12968 | |
| 100 μg | |
| Primary | |
| Detects human, mouse and rat NFATC3 in Western blots. | |
| Human, Mouse, Rat | |
| IgG |
| Western Blot | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| NFAT4, NF-AT4, NFAT4nuclear factor of activated T-cells, cytoplasmic 3, NFATc3, NF-ATc3, NFATx, nuclear factor of activated T-cells c3 isoform IE-Xa, nuclear factor of activated T-cells, cytoplasmic, calcineurin-dependent 3, T cell transcription factor NFAT4, T-cell transcription factor NFAT4 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 4775 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction