missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse/Rat Neurolysin Antibody, R&D Systems™
Sheep Polyclonal Antibody
Brand: R&D Systems AF3814
This item is not returnable.
View return policy
Description
Neurolysin Polyclonal specifically detects Neurolysin in Human, Mouse, Rat samples. It is validated for Western Blot, Immunoprecipitation.
Specifications
| Neurolysin | |
| Polyclonal | |
| Unconjugated | |
| AGTBP, AGTBPFLJ23002, angiotensin binding protein, Angiotensin-binding protein, DKFZp564F123, EC 3.4.24, EC 3.4.24.16, KIAA1226EP24.16, MEP, mitochondrial, MOP, neurolysin (metallopeptidase M3 family), Neurotensin Endopeptidase, NLN, Oligopeptidase M | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 57486 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Immunoprecipitation | |
| LYOPH | |
| Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL | |
| NLN | |
| E. coli-derived recombinant human Neurolysin aa 38-704 | |
| 100 μg | |
| Primary | |
| Detects human, mouse, and rat Neurolysin Neurolysin in Western blots and human Neurolysin in direct ELISAs. In direct ELISAs, less than 5% cross-reactivity with recombinant human (rh) Meprin alpha , rhMeprin beta subunit, and rhTHOP1 is observed. | |
| Human, Mouse, Rat | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction