missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse/Rat Neurolysin Antibody, R&D Systems™

Sheep Polyclonal Antibody
Brand: R&D Systems AF3814-SP
This item is not returnable.
View return policy
Description
Neurolysin Polyclonal specifically detects Neurolysin in Human, Mouse, Rat samples. It is validated for Western Blot, Immunoprecipitation.
Specifications
| Neurolysin | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| NLN | |
| E. coli-derived recombinant human Neurolysin aa 38-704 | |
| 25 μg | |
| Primary | |
| Detects human, mouse, and rat Neurolysin Neurolysin in Western blots and human Neurolysin in direct ELISAs. In direct ELISAs, less than 5% cross-reactivity with recombinant human (rh) Meprin alpha , rhMeprin beta subunit, and rhTHOP1 is observed. | |
| Human, Mouse, Rat | |
| IgG |
| Western Blot, Immunoprecipitation | |
| LYOPH | |
| Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL | |
| AGTBP, AGTBPFLJ23002, angiotensin binding protein, Angiotensin-binding protein, DKFZp564F123, EC 3.4.24, EC 3.4.24.16, KIAA1226EP24.16, MEP, mitochondrial, MOP, neurolysin (metallopeptidase M3 family), Neurotensin Endopeptidase, NLN, Oligopeptidase M | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 57486 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction