missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse/Rat Neurogranin Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB7947-SP
This item is not returnable.
View return policy
Description
Neurogranin Monoclonal specifically detects Neurogranin in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| Neurogranin | |
| Monoclonal | |
| LYOPH | |
| Western Blot 2 ug/mL, Immunohistochemistry 8-25 ug/mL | |
| Q92686 | |
| NRGN | |
| E. coli-derived recombinant human Neurogranin Met1-Asp78 Accession # Q92686 | |
| 25 μg | |
| Primary | |
| Detects human Neurogranin in ELISAs. | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Immunohistochemistry | |
| 898502 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in TBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| calmodulin-binding protein, hng, neurogranin, neurogranin (protein kinase C substrate, RC3), Ng, NRGN, protein kinase C substrate, RC3, RC3ng | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 4900 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG3 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction