missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse/Rat Neurofascin Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB3235-SP
This item is not returnable.
View return policy
Description
Neurofascin Monoclonal specifically detects Neurofascin in Human, Mouse, Rat samples. It is validated for Western Blot.
Specifications
| Neurofascin | |
| Monoclonal | |
| LYOPH | |
| Western Blot 0.5 ug/mL | |
| NP_446361 | |
| NFASC | |
| Mouse myeloma cell line NS0-derived recombinant rat Neurofascin Ile25-Ala1031 Accession # NP_446361 | |
| 25 μg | |
| Primary | |
| Detects rat Neurofascin in direct ELISAs and Western blots. | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot | |
| 727030 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| DKFZp686P2250, KIAA0756FLJ46866, neurofascin, neurofascin homolog, neurofascin homolog (chicken), NF, NFASC, NRCAML | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 23114 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction