missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse/Rat Netrin-1 Antibody, R&D Systems™

Sheep Polyclonal Antibody has been used in 4 publications
Brand: R&D Systems AF6419-SP
This item is not returnable.
View return policy
Description
Netrin-1 Polyclonal specifically detects Netrin-1 in Human, Mouse, Rat samples. It is validated for Western Blot.
Specifications
| Netrin-1 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| netrin 1, netrin-1, Netrin1, NTN1, NTN1Lnetrin 1, mouse, homolog of | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 9423 | |
| Sterile PBS to a final concentration of 0.2 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot | |
| LYOPH | |
| Western Blot 0.5 ug/mL | |
| O95631 | |
| NTN1 | |
| Mouse myeloma cell line NS0-derived recombinant human Netrin-1 Val22-Ala604 Accession # O95631 | |
| 25 μg | |
| Primary | |
| Detects human, mouse, and rat Netrin-1 in Western blots. | |
| Human, Mouse, Rat | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction