missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse/Rat NEDD8 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB4936-SP
This item is not returnable.
View return policy
Description
NEDD8 Monoclonal specifically detects NEDD8 in Human, Mouse, Rat samples. It is validated for Western Blot.
Specifications
| NEDD8 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 0.25 ug/mL | |
| Q15843 | |
| NEDD8 | |
| E. coli-derived recombinant human NEDD8 Leu2-Gly77 Accession # Q15843 | |
| 25 μg | |
| Primary | |
| Detects human, mouse, and rat NEDD8 in Western blots. | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot | |
| 584606 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| FLJ43224, MGC104393, MGC125896, MGC125897, NEDD-8, Neddylin, Neural precursor cell expressed developmentally down-regulated protein 8, neural precursor cell expressed, developmentally down-regulated 8, Rub1, Ubiquitin-like protein Nedd8 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 4738 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction