missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Human/Mouse/Rat Myostatin Recombinant Protein, PeproTech®

Product Code. 17861133
Click to view available options
Quantity:
1 mg
10 μg
100 μg
2 μg
250 μg
50 μg
500 μg
Unit Size:
100µg
10µg
1mg
250µg
2µg
500µg
50µg
This item is not returnable. View return policy

Product Code. 17861133

Brand: Gibco™ 12000250UG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

The amino acid sequence of mature myostatin is extremely conserved across species, and is the same in murine, rat, chicken, turkey, porcine, and human. Myostatin is expressed as the C-terminal part of a precursor polypeptide, which also contains a short N-terminal signal sequence for secretion, and a propeptide of 243 amino acids. After dimerization of this precursor, the covalent bonds between the propeptide and the mature ligand are cleaved by furin-type proteases. However, the resulting two proteins remain associated through non-covalent interactions, and are secreted as a latent complex. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

The protein encoded by this gene is a member of the bone morphogenetic protein family and the TGF-beta superfamily. This group of proteins is characterized by a polybasic proteolytic processing site which is cleaved to produce a mature protein containing seven conserved cysteine residues. The members of this family are regulators of cell growth and differentiation in both embryonic and adult tissues. This gene is thought to encode a secreted protein which negatively regulates skeletal muscle growth.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number O08689, O14793, O35312
For Use With (Application) Functional Assay
Formulation protein with no preservative
Gene ID (Entrez) 17700, 2660, 29152
Molecular Weight (g/mol) 25 kDa
Name Human/Mouse/Rat Myostatin
Quantity 250 μg
Source E. coli
Regulatory Status RUO
Endotoxin Concentration <1 EU/ μg
Gene Alias Cmpt; compact; Gdf8; GDF-8; growth differentiation factor 8; growth/differentiation factor 8; H-Myo-Propeptide; H-Myostatin; MSLHP; MSTN; Myostatin
Common Name Myostatin
Gene Symbol Mstn
Biological Activity Determined by its ability to inhibit the proliferation of MPC-11 cells. The expected ED50 for this effect is 17.0-25.0 ng/ml
Conjugate Unconjugated
Recombinant Recombinant
Sequence DFGLDCDEHS TESRCCRYPL TVDFEAFGWD WIIAPKRYKA NYCSGECEFV FLQKYPHTHL VHQANPRGSA GPCCTPTKMS PINMLYFNGK EQIIYGKIPA MVVDRCGCS
Content And Storage -20°C
Expression System E. coli
Form Lyophilized
Purity or Quality Grade ≥ 98% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.