missing translation for 'onlineSavingsMsg'
Learn More

Human/Mouse/Rat Muscle Phosphofructokinase/PFKM Antibody, R&D Systems™
GREENER_CHOICE

Product Code. 18049241
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18049241 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18049241 Supplier R&D Systems Supplier No. AF7687

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sheep Polyclonal Antibody

Muscle Phosphofructokinase/PFKM/PFK-1 Polyclonal specifically detects Muscle Phosphofructokinase/PFKM/PFK-1 in Human, Mouse, Rat samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Muscle Phosphofructokinase/PFKM/PFK-1
Applications Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.5 ug/mL
Gene Accession No. P08237
Gene Alias 6-phosphofructokinase, muscle type, EC 2.7.1,6-phosphofructo-1-kinase, EC 2.7.1.11, GSD7, MGC8699, PFK1, PFK-1, PFKA, PFK-A, PFKM, PFKX, Phosphofructo-1-kinase isozyme A, Phosphofructokinase 1, phosphofructokinase, muscle, phosphofructokinase, polypeptide X, Phosphofructokinase-1, Phosphofructokinase-M, phosphohexokinase
Gene Symbols PFKM
Host Species Sheep
Immunogen E. coli-derived recombinant human Phosphofructokinase/PFKM Asn674-Val780 Accession # P08237
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5213
Test Specificity Detects human, mouse, and rat Phosphofructokinase/PFKM in direct ELISAs and Western blots.
Reconstitution Sterile PBS to a final concentration of 0.2 mg/mL.
Target Species Human, Mouse, Rat
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.