missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse/Rat Muscle Phosphofructokinase/PFKM Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB7687-SP
This item is not returnable.
View return policy
Description
Muscle Phosphofructokinase/PFKM/PFK-1 Monoclonal specifically detects Muscle Phosphofructokinase/PFKM/PFK-1 in Human, Mouse, Rat samples. It is validated for Western Blot, Simple Western.
Specifications
| Muscle Phosphofructokinase/PFKM/PFK-1 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 0.2 ug/mL, Simple Western 2 ug/mL | |
| P08237 | |
| PFKM | |
| E. coli-derived recombinant human Muscle Phosphofructokinase/PFKM Asn674-Val780 Accession # P08237 | |
| 25 μg | |
| Primary | |
| Detects human Muscle Phosphofructokinase/PFKM in direct ELISAs and Western blots. Detects mouse and rat Muscle Phosphofructokinase/PFKM in Western blots. | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot | |
| 842735 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| 6-phosphofructokinase, muscle type, EC 2.7.1,6-phosphofructo-1-kinase, EC 2.7.1.11, GSD7, MGC8699, PFK1, PFK-1, PFKA, PFK-A, PFKM, PFKX, Phosphofructo-1-kinase isozyme A, Phosphofructokinase 1, phosphofructokinase, muscle, phosphofructokinase, polypeptide X, Phosphofructokinase-1, Phosphofructokinase-M, phosphohexokinase | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 5213 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction