missing translation for 'onlineSavingsMsg'
Learn More

Human/Mouse/Rat MLK4 alpha Antibody, R&D Systems™
GREENER_CHOICE

Product Code. 18021772
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18021772 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18021772 Supplier R&D Systems Supplier No. AF3435

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

MLK4 alpha Polyclonal specifically detects MLK4 alpha in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MLK4 alpha
Applications Western Blot, Immunohistochemistry
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Immunohistochemistry 5-15 ug/mL
Gene Accession No. CAC84639
Gene Alias dJ862P8.3, KIAA1804, MAP3K21, MLK4
Gene Symbols KIAA1804
Host Species Rabbit
Immunogen Human MLK4 alpha synthetic peptide (aa 560 - 570) ELSALPPGLLC Accession # CAC84639
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84451
Test Specificity Detects endogenous human, mouse, and rat MLK4 alpha . Because the peptide immunogen corresponds to a region at the C-terminus of MLK4 alpha absent in MLK4 beta , no reactivity with MLK4 beta is present.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human, Mouse, Rat
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.