missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse/Rat MKP-3 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB3576-SP
This item is not returnable.
View return policy
Description
MKP-3/DUSP6 Monoclonal specifically detects MKP-3/DUSP6 in Human, Mouse, Rat samples. It is validated for Western Blot.
Specifications
| MKP-3/DUSP6 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| Q16828 | |
| DUSP6 | |
| E. coli-derived recombinant human MKP-3 Ile2-Thr381 Accession # Q16828 | |
| 25 μg | |
| Primary | |
| Detects endogenous human, mouse, and rat MKP-3 in Western blots. In Western blots, this antibody does not cross-react with recombinant human MKP-1 or MKP-2. | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot | |
| 363246 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| dual specificity phosphatase 6, Dual specificity protein phosphatase PYST1, DUSP6, EC 3.1.3.16, EC 3.1.3.48, MAP kinase phosphatase 3, Mitogen-activated protein kinase phosphatase 3, MKP3, MKP-3, MKP3serine/threonine specific protein phosphatase, PYST1, PYST1dual specificity protein phosphatase 6 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 1848 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction