missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse/Rat MKK3 Antibody, R&D Systems™
Mouse Monoclonal Antibody
Brand: R&D Systems MAB2515
This item is not returnable.
View return policy
Description
MKK3/MEK3 Monoclonal specifically detects MKK3/MEK3 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry.
Specifications
| MKK3/MEK3 | |
| Monoclonal | |
| Unconjugated | |
| Supplied as a 0.2 μm filtered solution in PBS. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| MAP kinase kinase 3, MAP2K3, MAPK/ERK kinase 3, MAPKK 3, MAPKK3PRKMK3dual specificity mitogen-activated protein kinase kinase 3, MEK3, MEK3MEK 3, mitogen activated protein kinase kinase 3, mitogen-activated protein kinase kinase 3, MKK3EC 2.7.12.2, PRKMK3 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 5606 | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Immunocytochemistry | |
| 275909 | |
| Western Blot 1 ug/mL, Immunocytochemistry 15-30 ug/mL | |
| P46734 | |
| MAP2K3 | |
| E. coli-derived recombinant human MKK3 Met1-Ser347 Accession # P46734 | |
| 100 μg | |
| Primary | |
| Detects human MKK3 in direct ELISAs and Western blots. In Western blots, no cross-reactivity with recombinant human MKK6 is observed. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC, as supplied. 1 month, 2 to 8 degreesC under sterile conditions after opening. 6 months, -20 to -70 degreesC under sterile conditions after opening. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction