missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse/Rat MEKK2 Antibody, R&D Systems™
Mouse Monoclonal Antibody
Brand: R&D Systems MAB7128
This item is not returnable.
View return policy
Description
MEKK2 Monoclonal specifically detects MEKK2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry.
Specifications
| MEKK2 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 2 ug/mL, Immunocytochemistry 8-25 ug/mL | |
| Q9Y2U5 | |
| MAP3K2 | |
| E. coli-derived recombinant human MEKK2 Asn42-Tyr264 Accession # Q9Y2U5 | |
| 100 μg | |
| Primary | |
| Detects recombinant human MEKK2 in direct ELISA and human, mouse, rat MEKK2 in Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human MEKK1, 3, or 4 is observed. | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Immunocytochemistry | |
| 722728 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| EC 2.7.11.25, MAP/ERK kinase kinase 2, MAP3K2, MAPK/ERK kinase kinase 2, MAPKKK2, MEK kinase 2, MEKK 2, MEKK2, MEKK2B, mitogen-activated protein kinase kinase kinase 2 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 10746 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction