missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse/Rat Lipin 2 Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems AF3756-SP
This item is not returnable.
View return policy
Description
Lipin 2 Polyclonal specifically detects Lipin 2 in Human, Mouse, Rat samples. It is validated for Western Blot.
Specifications
| Lipin 2 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| EC 3.1.3.4, KIAA0249lipin-2, lipin 2, Lipin-2, LPIN2, phosphatidate phosphatase LPIN2 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 9663 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot | |
| LYOPH | |
| Western Blot 0.1 ug/mL | |
| Q92539 | |
| LPIN2 | |
| E. coli-derived recombinant human Lipin 2 Ser439-Lys563 Accession # Q92539 | |
| 25 μg | |
| Primary | |
| Detects Lipin 2 in direct ELISAs and Western blots. In direct ELISAs and Western blots, less than 1% cross-reactivity with recombinant human (rh) Lipin 1 and rhLipin 3 is observed. | |
| Human, Mouse, Rat | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction