missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse/Rat Lipin 2 Antibody, R&D Systems™
Goat Polyclonal Antibody has been used in 1 publication
Brand: R&D Systems AF3756
Additional Details : Weight : 0.00970kg
Description
Lipin 2 Polyclonal specifically detects Lipin 2 in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
Lipin 2 | |
Polyclonal | |
Unconjugated | |
Q92539 | |
LPIN2 | |
E. coli-derived recombinant human Lipin 2 Ser439-Lys563 Accession # Q92539 | |
100 μg | |
Primary | |
Detects Lipin 2 in direct ELISAs and Western blots. In direct ELISAs and Western blots, less than 1% cross-reactivity with recombinant human (rh) Lipin 1 and rhLipin 3 is observed. | |
Human, Mouse, Rat | |
IgG |
Western Blot | |
LYOPH | |
Western Blot 0.1 ug/mL | |
EC 3.1.3.4, KIAA0249lipin-2, lipin 2, Lipin-2, LPIN2, phosphatidate phosphatase LPIN2 | |
Goat | |
Affinity Purified | |
RUO | |
9663 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only