missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse/Rat JNK Pan Specific Antibody, R&D Systems™

Rabbit Polyclonal Antibody has been used in 9 publications
Brand: R&D Systems AF1387-SP
This item is not returnable.
View return policy
Description
JNK1/2/3 Polyclonal specifically detects JNK1/2/3 in Human, Mouse, Rat samples. It is validated for Western Blot, Simple Western, Immunohistochemistry.
Specifications
| JNK1/2/3 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| c-Jun N-terminal kinase 1, EC 2.7.11, EC 2.7.11.24, JNK1 alpha protein kinase, JNK1 beta protein kinase, JNK1JNK1A2, JNK-46, JUN N-terminal kinase, MAP kinase 8, MAPK 8, MAPK10, MAPK8, MAPK9, mitogen-activated protein kinase 8, mitogen-activated protein kinase 8 isoform JNK1 alpha1, mitogen-activated protein kinase 8 isoform JNK1 beta2, PRKM8, PRKM8JNK, protein kinase JNK1, SAPK1, SAPK1 alpha, SAPK1 beta, SAPK1JNK21B1/2, Stress-activated protein kinase 1, Stress-activated protein kinase JNK1 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 5599 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Immunohistochemistry | |
| LYOPH | |
| Western Blot 0.2 ug/mL, Simple Western 10 ug/mL, Immunohistochemistry 5-15 ug/mL | |
| P45983 | |
| MAPK8IP3 | |
| E. coli-derived recombinant human JNK1 Met1-Gln384 Accession # P45983 | |
| 25 μg | |
| Primary | |
| Detects human, mouse, and rat p46 and p54 JNK in Western blots. Detects recombinant human (rh) JNK1, rhJNK2, and rhJNK3. | |
| Human, Mouse, Rat | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction