missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse/Rat Jak1 Alexa Fluor 647-conjugated Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems IC4260R-100UG
This item is not returnable.
View return policy
Description
Jak1 Monoclonal specifically detects Jak1 in Human, Mouse, Rat samples. It is validated for Intracellular Staining by Flow Cytometry.
Specifications
| Jak1 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells | |
| P23458 | |
| JAK1 | |
| E. coli-derived recombinant human Jak1 Pro32-Phe286 Accession # P23458 | |
| RUO | |
| 3716 | |
| Human, Mouse, Rat | |
| IgG2b |
| Flow Cytometry | |
| 413104 | |
| Alexa Fluor 647 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| EC 2.7.10, JAK-1, JAK1Atyrosine-protein kinase JAK1, JAK1B, Janus kinase 1EC 2.7.10.2, JTK3 | |
| Rat | |
| 100 μg | |
| Primary | |
| Detects human, mouse, and rat Jak1. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction