missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Human/Mouse/Rat Irisin Recombinant Protein, PeproTech®

Product Code. 17839892
Click to view available options
Quantity:
1 mg
10 μg
100 μg
2 μg
250 μg
50 μg
500 μg
Unit Size:
100µg
10µg
1mg
250µg
2µg
500µg
50µg
This item is not returnable. View return policy

Product Code. 17839892

Brand: Gibco™ 10065100UG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Recombinant Human/Murine/Rat Irisin, appears to form a non-disulfide linked oligomeric structure in solution, has a calculated theoretical molecular weight of 25.2 kDa, but migrates at an apparent molecular weight of 28 kDa by SDS-PAGE analysis under reducing conditions due to glycosylation. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

Fibronectin type III domain-containing protein 5, the precursor of irisin, is a protein that is encoded by the FNDC5 gene. This gene encodes a secreted protein that is released from muscle cells during exercise. The encoded protein may participate in the development of brown fat. Translation of the precursor protein initiates at a non-AUG start codon at a position that is conserved as an AUG start codon in other organisms. Alternative splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q8K3V5, Q8K4Z2, Q8NAU1
For Use With (Application) Functional Assay
Formulation protein with no preservative
Gene ID (Entrez) 252995, 260327, 384061
Molecular Weight (g/mol) 25.2 kDa
Name Human/Mouse/Rat Irisin
Quantity 100 μg
Source CHO cells
Regulatory Status RUO
Endotoxin Concentration <1 EU/ μg
Gene Alias 1500001L03Rik; AI836596; C87088; fibronectin type III domain containing 5; fibronectin type III domain-containing protein 5; Fibronectin type III repeat-containing protein 2; Fndc5; Frcp2; Irisin; PeP; Peroxisomal protein; Pxp
Common Name FNDC5
Gene Symbol FNDC5
Biological Activity Determined by its ability to promote cell survival and growth of rat H9c2 cardio-myoblasts under serum starvation conditions.
Conjugate Unconjugated
Recombinant Recombinant
Sequence DSPSAPVNVT VRHLKANSAV VSWDVLEDEV VIGFAISQQK KDVRMLRFIQ EVNTTTRSCA LWDLEEDTEY IVHVQAISIQ GQSPASEPVL FKTPREAEKM ASKNKDEVTM KE
Content And Storage -20°C
Expression System CHO cells
Form Lyophilized
Purity or Quality Grade ≥ 90% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.