missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse/Rat Inosine 5'-Monophosphate Dehydrogenase 2/IMPDH2 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB8349-SP
This item is not returnable.
View return policy
Description
IMP Dehydrogenase 2/IMPDH2 Monoclonal specifically detects IMP Dehydrogenase 2/IMPDH2 in Human, Mouse, Rat samples. It is validated for Western Blot, Simple Western.
Specifications
| IMP Dehydrogenase 2/IMPDH2 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| EC 1.1.1.205, IMDH2, IMP (inosine 5'-monophosphate) dehydrogenase 2, IMP (inosine monophosphate) dehydrogenase 2, IMP dehydrogenase 2, IMP Oxireductase 2, IMPD 2, IMPD2, IMPD2IMP oxireductase 2, IMPDH 2, IMPDH2, IMPDH-II, inosine 5' phosphate dehydrogenase 2, inosine monophosphate dehydrogenase type II, inosine-5'-monophosphate dehydrogenase 2 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 3615 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
| Western Blot | |
| 911425 | |
| Western Blot 0.5 ug/mL, Simple Western 5 ug/mL | |
| P12268 | |
| IMPDH2 | |
| S. frugiperda insect ovarian cell line Sf 21-derived recombinant human Inosine 5'-Monophosphate Dehydrogenase 2/IMPDH2 Met1-Phe514 Accession # P12268 | |
| 25 μg | |
| Primary | |
| Detects human Inosine 5'-Monophosphate Dehydrogenase 2/IMPDH2 in direct ELISAs. Detects human, mouse and rat IMPDH2 in Western blots. | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction