missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse/Rat Importin beta/KPNB1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB8209
This item is not returnable.
View return policy
Description
Importin beta/KPNB1 Monoclonal specifically detects Importin beta/KPNB1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry.
Specifications
| Importin beta/KPNB1 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL, Immunocytochemistry 8-25 ug/mL | |
| IMB1, Impnb, importin 1, importin 90, importin beta-1 subunit, importin subunit beta-1, importin-90, IPO1, IPOB, karyopherin (importin) beta 1, Karyopherin subunit beta-1, KPNB1, MGC2155, MGC2156, MGC2157, NTF97, NTF97Pore targeting complex 97 kDa subunit, Nuclear factor p97, PTAC97 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| Primary | |
| Detects human Importin beta/KPNB1 in ELISA and Western Blot. It detects mouse and rat Importin beta/KPNB1 in Western Blots. | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Immunocytochemistry | |
| 845208 | |
| Unconjugated | |
| Q14974 | |
| KPNB1 | |
| E. coli-derived recombinant human Importin beta/KPNB1 Met1-Gly155 Accession # Q14974 | |
| 100 μg | |
| 3837 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction